missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Valine methyl ester hydrochloride, 99%
CAS: 5619-05-6 | C9H12O2 | 152.19 g/mol
$86.34 - $330.78
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAL0652606
|
N/A | 5 g |
Each for $86.34
|
|
|||||
|
AAL0652614
|
N/A | 25 g |
Each for $330.78
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5619-05-6 | |
| MFCD00066118 |
Specifications
| 112°C to 114°C | |
| C9H12O2 | |
| 3912090 | |
| FPEUDBGJAVKAEE-UHFFFAOYSA-N | |
| 1,3-dimethoxy-2-methylbenzene | |
| 79755 | |
| 99% |
| 5619-05-6 | |
| MFCD00066118 | |
| 2,6-dimethoxytoluene, benzene, 1,3-dimethoxy-2-methyl, 2-methyl-1,3-bis methyloxy benzene, 1,3-dimethoxy-2-methylbenzol, pubchem4545, acmc-1aqu0, 3-chloro-6-hydroxybiphenyl, 1,3-dimethoxy-2-methyl-benzene, # | |
| CC1=C(C=CC=C1OC)OC | |
| 152.19 | |
| 167.64 | |
| DL-Valine methyl ester hydrochloride |
Safety and Handling
EINECSNumber : 227-048-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only