accessibility menu, dialog, popup

UserName

CH3OONH2H3CCH3HCl

CAS RN 5619-05-6

IUPAC Name: methyl 2-amino-3-methylbutanoate hydrochloride
Synonyms: methyl 2-amino-3-methylbutanoate hydrochloride
Molecular Weight (g/mol): 167.63
Molecular Formula: C6H14ClNO2
InChi Key: KUGLDBMQKZTXPW-UHFFFAOYSA-N
SMILES: Cl.COC(=O)C(N)C(C)C
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)