missing translation for 'onlineSavingsMsg'
Learn More

DL-Valine methyl ester hydrochloride, 99%

Catalog No. AAL0652606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0652606 5 g
AAL0652614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0652606 Supplier Thermo Scientific Chemicals Supplier No. L0652606

CAS: 5619-05-6 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5619-05-6
MDL Number MFCD00066118

Specifications

Melting Point 112°C to 114°C
Molecular Formula C9H12O2
Beilstein 3912090
Synonym 2,6-dimethoxytoluene, benzene, 1,3-dimethoxy-2-methyl, 2-methyl-1,3-bis methyloxy benzene, 1,3-dimethoxy-2-methylbenzol, pubchem4545, acmc-1aqu0, 3-chloro-6-hydroxybiphenyl, 1,3-dimethoxy-2-methyl-benzene, #
InChI Key FPEUDBGJAVKAEE-UHFFFAOYSA-N
SMILES CC1=C(C=CC=C1OC)OC
IUPAC Name 1,3-dimethoxy-2-methylbenzene
Quantity 5 g
Molecular Weight (g/mol) 152.19
PubChem CID 79755
Formula Weight 167.64
Percent Purity 99%
Chemical Name or Material DL-Valine methyl ester hydrochloride
Show More Show Less
EINECSNumber 227-048-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only