missing translation for 'onlineSavingsMsg'
Learn More

DL-Valine methyl ester hydrochloride, 99%

Catalog No. AAL0652614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0652614 25 g
AAL0652606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0652614 Supplier Thermo Scientific Chemicals Supplier No. L0652614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5619-05-6 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5619-05-6
MDL Number MFCD00066118

Specifications

Melting Point 112°C to 114°C
Molecular Formula C6H14ClNO2
Beilstein 3912090
Synonym dl-valine methyl ester hydrochloride, h-dl-val-ome.hcl, methyl 2-amino-3-methylbutanoate hydrochloride, h-d-val-ome.hcl, methyl dl-valinate hydrochloride, d,l-valine methyl ester hydrochloride, h-d-val-ome*hcl, h-val-ome??hcl, acmc-209fna, acmc-209ncl
InChI Key KUGLDBMQKZTXPW-UHFFFAOYSA-N
SMILES Cl.COC(=O)C(N)C(C)C
IUPAC Name methyl 2-amino-3-methylbutanoate;hydrochloride
Molecular Weight (g/mol) 167.63
Quantity 25 g
PubChem CID 12498813
Formula Weight 167.64
Percent Purity 99%
Chemical Name or Material DL-Valine methyl ester hydrochloride
Show More Show Less
EINECSNumber 227-048-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only