missing translation for 'onlineSavingsMsg'
Learn More

DL-Tropic acid, 98%

Catalog No. AAB2204018
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2204018 50 g
AAB2204009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2204018 Supplier Thermo Scientific Chemicals Supplier No. B2204018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 552-63-6 | C9H10O3 | 166.18 g/mol

It is a laboratory reagent used in the chemical synthesis of atropine and hyoscyamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a laboratory reagent used in the chemical synthesis of atropine and hyoscyamine.

Solubility
Soluble in water (20 g/L) at 20°C, ethanol, ether, alcohol, and methanol (0.1 g/ml).

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 552-63-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004255
InChI Key JACRWUWPXAESPB-UHFFFAOYNA-N
Synonym tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid
PubChem CID 10726
ChEBI CHEBI:30765
IUPAC Name 3-hydroxy-2-phenylpropanoic acid
SMILES OCC(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 117°C to 119°C
Quantity 50 g
Beilstein 2209199
Merck Index 14,9779
Solubility Information Soluble in water (20g/L) at 20°C,ethanol,ether,alcohol,and methanol (0.1g/ml).
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material DL-Tropic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-020-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only