Learn More
Abcam Inc D,L-Thioctic acid (DL-alpha-Lipoic acid), Non-specific free radical scavenger, 5G
Numéro de catalogue. 502706145
Encompass
MW 206.3 Da, Purity >98%. Non-specific free radical scavenger. Prevents lipid peroxidation and promotes mitochondrial permeability transition in vitro. Anti-oxidant. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502706145 Fournisseur Abcam Inc Code fournisseur AB1429525G
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
MW 206.3 Da, Purity >98%. Non-specific free radical scavenger. Prevents lipid peroxidation and promotes mitochondrial permeability transition in vitro. Anti-oxidant. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement