missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Tartaric Acid 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 199°C |
| Color | White |
| Quantity | 500 g |
| Physical Form | Crystalline Powder |
| Formula Weight | 150.09 |
| Percent Purity | ≥99.0% (T) |
| Chemical Name or Material | DL-Tartaric Acid |
Safety and Handling
| EINECSNumber | (2)-1456 |
| TSCA | Yes |