missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Proline, 99%

Catalog No. AC174030250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC174030250 25 g Glass Bottle
AC174030050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC174030250 Supplier Thermo Scientific Chemicals Supplier No. 174030250
Only null left

CAS: 609-36-9 | C5H9NO2 | 115.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 609-36-9
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00005250
InChI Key ONIBWKKTOPOVIA-UHFFFAOYSA-N
Synonym dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275
PubChem CID 614
ChEBI CHEBI:26271
IUPAC Name pyrrolidine-2-carboxylic acid
SMILES C1CC(NC1)C(=O)O

Specifications

Melting Point 208.0°C to 210.0°C
Color White
Heavy Metals (as Pb) 20ppm max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Residue after Ignition 0.1% max.
Beilstein 22, 4
Fieser 09,393
Merck Index 15, 7895
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohol, sparingly soluble in chloroform, benzene and, acetone, insoluble in ether
Water 0.5% max. (K.F.)
Quantity 25 g
Formula Weight 115.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material DL-Proline, 99%
Show More Show Less
EINECSNumber 210-189-3
TSCA TSCA

RUO – Research Use Only