missing translation for 'onlineSavingsMsg'
Learn More

DL-Phenylalanine 98.0+%, TCI America™

Catalog No. P013625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P013625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P013625G Supplier TCI America Supplier No. P013625G

Specifications

CAS 150-30-1
Color White
Molecular Formula C9H11NO2
Quantity 25 g
MDL Number MFCD00064225
Synonym dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
SMILES NC(CC1=CC=CC=C1)C(O)=O
IUPAC Name 2-amino-3-phenylpropanoic acid
Molecular Weight (g/mol) 165.19
PubChem CID 994
ChEBI CHEBI:28044
Formula Weight 165.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Phenylalanine
Show More Show Less
EINECSNumber (9)-1283
TSCA Yes