missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Phenylalanine, 99%

Catalog No. AC164575000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164575000 500 g Plastic Bottle
AC164570250 25 g Plastic Bottle
AC164571000 100 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC164575000 Supplier Thermo Scientific Chemicals Supplier No. 164575000

DL-Phenylalanine, 99%, C9H11NO2, CAS Number-150-30-1, 2-amino-3-phenylpropionic acid, alanine, phenyl-, dl, dl-2-amino-3-phenylpropanoic acid, dl-3-phenylalanine, dlpa, dl-phenylalanine, h-dl-phe-oh, phenylalanin, phenylalanine, phenylalanine, dl, 100g, 14,498, CHEBI:28044, White, 205-756-7, 165.19 | CAS: 150-30-1 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 150-30-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00064225
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
PubChem CID 994
ChEBI CHEBI:28044
IUPAC Name 2-amino-3-phenylpropanoic acid
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 266.0°C
Color White
Loss on Drying 0.3% (105°C, 2 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic Bottle
Linear Formula C6H5CH2CH(NH2)CO2H
Quantity 500 g
Beilstein 14,498
Merck Index 15,7382
Solubility Information Solubility in water: 14.11g/L (25°C)
Formula Weight 165.19
Percent Purity 99%
Physical Form Powder
Chemical Name or Material DL-Phenylalanine
Show More Show Less
EINECSNumber 205-756-7

RUO – Research Use Only