missing translation for 'onlineSavingsMsg'
Learn More

DL-Panthenol, 99%

Catalog No. AAB2511430
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2511430 250 g
AAB2511418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2511430 Supplier Thermo Scientific Chemicals Supplier No. B2511430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16485-10-2 | C9H19NO4 | 205.25 g/mol

DL-Panthenol is used in enantiospecific HPLC separation of pantothenic acid, panthenol and pantolactone enantiomers. It promotes stimulation of epithelization, granulation, mitigation of itching and has an anti-inflammatory effect. It is effective for moisturization, improvement of hair structure and adding luster and sheen to the hair.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Panthenol is used in enantiospecific HPLC separation of pantothenic acid, panthenol and pantolactone enantiomers. It promotes stimulation of epithelization, granulation, mitigation of itching and has an anti-inflammatory effect. It is effective for moisturization, improvement of hair structure and adding luster and sheen to the hair.

Solubility
Soluble in water, ethanol, ether, chloroform, and propylene glycol.

Notes
Hygroscopic. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.

Chemical Identifiers

CAS 16485-10-2
Molecular Formula C9H19NO4
Molecular Weight (g/mol) 205.25
MDL Number MFCD00002944
InChI Key SNPLKNRPJHDVJA-UHFFFAOYNA-N
Synonym dl-panthenol, panthenol, dl-pantothenol, dl-pantothenyl alcohol, panthenolum, 2,4-dihydroxy-n-3-hydroxypropyl-3,3-dimethylbutanamide, pantenol, pantenolo, penthenol, varitan
PubChem CID 4678
IUPAC Name 2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
SMILES CC(C)(CO)C(O)C(=O)NCCCO

Specifications

Melting Point 66°C to 69°C
Quantity 250 g
Sensitivity Hygroscopic
Merck Index 14,2947
Solubility Information Soluble in water,ethanol,ether,chloroform,and propylene glycol.
Formula Weight 205.26
Percent Purity 99%
Chemical Name or Material DL-Panthenol
EINECSNumber 240-540-6
RTECSNumber ES4316500
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only