missing translation for 'onlineSavingsMsg'
Learn More

DL-Norphenylephrine Hydrochloride 98.0+%, TCI America™

Catalog No. n038925g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
N038925G 25 g
N03895G 5 g
2 options

Catalog No. N038925G

Supplier: TCI America N038925G

Only null left

Chemical Identifiers

CAS 4779-94-6
Molecular Formula C8H12ClNO2
Molecular Weight (g/mol) 189.639
MDL Number MFCD00012879
InChI Key OWMFSWZUAWKDRR-UHFFFAOYSA-N
Synonym 3-(2-Amino-1-hydroxyethyl)phenol Hydrochloride
PubChem CID 3016408
IUPAC Name 3-(2-amino-1-hydroxyethyl)phenol;hydrochloride
SMILES C1=CC(=CC(=C1)O)C(CN)O.Cl

Specifications

Melting Point 157°C
Color White
Quantity 25 g
Formula Weight 189.64
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Norphenylephrine Hydrochloride
RTECSNumber DN7360000
TSCA No