missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Lysine Monohydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 182.65 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | DL-Lysine Monohydrochloride |
Safety and Handling
| TSCA | Yes |