missing translation for 'onlineSavingsMsg'
Learn More

DL-Leucine 98.0+%, TCI America™

Catalog No. L002825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L002825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L002825G Supplier TCI America Supplier No. L002825G
Only null left

Chemical Identifiers

CAS 328-39-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00063087
InChI Key ROHFNLRQFUQHCH-UHFFFAOYNA-N
Synonym dl-leucine, h-dl-leu-oh, leucine, dl, rs-leucine, +--leucine, dl-2-amino-4-methylpentanoic acid, 2-amino-4-methyl-pentanoic acid, l-1-13c leucine, l-1-14c leucine, leucine,d
PubChem CID 857
ChEBI CHEBI:25017
IUPAC Name 2-amino-4-methylpentanoic acid
SMILES CC(C)CC(N)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 131.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Leucine
EINECSNumber (9)-1632
TSCA Yes