missing translation for 'onlineSavingsMsg'
Learn More

DL-Isoleucine, 99%

Numéro de catalogue. AAA1752114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1752114 25 g
AAA1752122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAA1752114 Fournisseur Thermo Scientific Chemicals Code fournisseur A1752114
Il en reste null

CAS: 443-79-8 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 443-79-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00004268
InChI Key AGPKZVBTJJNPAG-WHFBIAKZSA-N
PubChem CID 94206
ChEBI CHEBI:20899
SMILES CC[C@H](C)[C@H](N)C(O)=O

Spécifications

Melting Point ∼280°C (decomposition)
Beilstein 1721796
Merck Index 14,5179
Synonym d-alloisoleucine, d-allo-isoleucine, 2r,3s-2-amino-3-methylpentanoic acid, allo-d-isoleucine, alloisoleucine, d, h-d-allo-ile-oh, threo-d-isoleucine, dl-allo-isoleucine, hile, h-dl-allo-ile-oh
IUPAC Name (2R,3S)-2-amino-3-methylpentanoic acid
Quantity 25 g
Formula Weight 131.18
Percent Purity 99%
Chemical Name or Material DL-Isoleucine
EINECSNumber 207-139-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only