missing translation for 'onlineSavingsMsg'
Learn More

DL-Glutamine 98.0+%, TCI America™

Catalog No. G00621G
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G00621G 1 g
G006210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G00621G Supplier TCI America Supplier No. G00621G
Only null left

Chemical Identifiers

CAS 6899-04-3
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00065103
InChI Key ZDXPYRJPNDTMRX-UHFFFAOYNA-N
Synonym H-DL-Gln-OH
PubChem CID 738
ChEBI CHEBI:28300
IUPAC Name 2,5-diamino-5-oxopentanoic acid
SMILES C(CC(=O)N)C(C(=O)O)N

Specifications

Color White
Quantity 1 g
Formula Weight 146.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Glutamine