missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Glutamic acid monohydrate, 98+%

Catalog No. AC119931000
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC119931000 100 g Plastic bottle
AC119930250 25 g Plastic bottle
AC119930010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC119931000 Supplier Thermo Scientific Chemicals Supplier No. 119931000

CAS: 19285-83-7 | C5H9NO4·H2O | 165.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19285-83-7
Molecular Formula C5H9NO4·H2O
Molecular Weight (g/mol) 165.15
MDL Number MFCD00150703
InChI Key OZDAOHVKBFBBMZ-UHFFFAOYSA-N
Synonym dl-glutamic acid monohydrate, 2-aminopentanedioic acid hydrate, dl-glutamic acid hydrate, h-dl-glu-oh.h2o, glutaminsaeure hydrate, +/--2-aminoglutaric acid, dl-2-aminopentanedioic acid, 2-azanylpentanedioic acid hydrate, 2-aminopentanedioic acid, oxamethane, hydrate, a-aminoglutarate
PubChem CID 16219385
IUPAC Name 2-aminopentanedioic acid;hydrate
SMILES C(CC(=O)O)C(C(=O)O)N.O

Specifications

Melting Point 194.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula HO2CCH2CH2CH(NH2)CO2H·H2O
Quantity 100 g
Solubility Information Solubility in water: 20.54g/L (25°C). Other solubilities: sparingly soluble in alcohol and ether
Formula Weight 165.15
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material DL-Glutamic acid monohydrate, 99%
Show More Show Less

RUO – Research Use Only