missing translation for 'onlineSavingsMsg'
Learn More

DL-Carnitine hydrochloride, 98+%

Catalog No. AAA1658922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1658922 100 g
AAA1658914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1658922 Supplier Thermo Scientific Chemicals Supplier No. A1658922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 461-05-2 | C7H17ClNO3+ | 198.667 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 461-05-2
Molecular Formula C7H17ClNO3+
Molecular Weight (g/mol) 198.667
MDL Number MFCD00011904
InChI Key JXXCENBLGFBQJM-UHFFFAOYSA-O
Synonym dl-carnitine hydrochloride, 3-carboxy-2-hydroxypropyl-trimethylammonium hydrochloride, 2,4-bis oxidanyl-4-oxidanylidene-butyl-trimethyl-azanium hydrochloride
PubChem CID 24206429
IUPAC Name (3-carboxy-2-hydroxypropyl)-trimethylazanium;hydrochloride
SMILES C[N+](C)(C)CC(CC(=O)O)O.Cl

Specifications

Melting Point 196°C to 200°C
Quantity 100 g
Beilstein 4163212
Sensitivity Hygroscopic
Merck Index 14,1849
Formula Weight 197.66
Percent Purity ≥98%
Chemical Name or Material DL-Carnitine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-309-1
RTECSNumber BP2979140
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only