missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Aspartic acid, 98+%

Catalog No. AAA1364636
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1364636 500 g
AAA1364622 100 g
AAA136460E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1364636 Supplier Thermo Scientific Chemicals Supplier No. A1364636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 617-45-8 | C4H7NO4 | 133.10 g/mol

DL-Aspartic acid is used in studies on factors and conditions that enhance semen and sperm quality. Its conjugate base D-aspartate has potential use as a therapeutic agent in the treatment of schizophrenia-related symptoms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Aspartic acid is used in studies on factors and conditions that enhance semen and sperm quality. Its conjugate base D-aspartate has potential use as a therapeutic agent in the treatment of schizophrenia-related symptoms.

Solubility
Soluble in water, and hydrochloric acid. Insoluble in ether, and alcohol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 617-45-8
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.10
MDL Number MFCD00063083
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
Synonym dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart
PubChem CID 424
ChEBI CHEBI:22660
IUPAC Name 2-aminobutanedioic acid
SMILES NC(CC(O)=O)C(O)=O

Specifications

Melting Point >300°C (decomposition)
Beilstein 774618
Solubility Information Soluble in water,and hydrochloric acid. Insoluble in ether,and alcohol.
Quantity 500 g
Formula Weight 133.1
Percent Purity ≥98%
Chemical Name or Material DL-Aspartic acid
EINECSNumber 210-513-3
RTECSNumber CI9097800
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only