missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Asparagine monohydrate, 98%

Catalog No. AAB2027336
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2027336 500 g
AAB2027322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2027336 Supplier Thermo Scientific Chemicals Supplier No. B2027336

CAS: 69833-18-7 | C4H8N2O3 | 132.12 g/mol

DL-Asparagine monohydrate is used as a pharmaceutical intermediate. It is used to prepare DL-asparaginium nitrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Asparagine monohydrate is used as a pharmaceutical intermediate. It is used to prepare DL-asparaginium nitrate.

Solubility
Soluble in water, acids, and alkalis.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 69833-18-7
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.12
MDL Number MFCD00151039,MFCD00008036,MFCD00064400
InChI Key DCXYFEDJOCDNAF-UHFFFAOYNA-N
Synonym dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina
PubChem CID 522362
SMILES NC(CC(N)=O)C(O)=O

Specifications

Melting Point ∼220°C (decomposition)
Beilstein 6234992
Solubility Information Soluble in water,acids,and alkalis.
IUPAC Name 2,4-diamino-4-oxobutanoic acid;hydrate
Quantity 500 g
Formula Weight 150.14 (132.12 Anhydrous)
Percent Purity 98%
Chemical Name or Material DL-Asparagine monohydrate
EINECSNumber 221-521-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only