missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Arginine, 98%

Catalog No. AAA1752014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1752014 25 g
AAA1752006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1752014 Supplier Thermo Scientific Chemicals Supplier No. A1752014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7200-25-1 | C6H14N4O2 | 174.204 g/mol

DL-Arginine is used in the synthesis of creatine and polyamines. DL-Arg is used in physicochemical analysis of amino acid complexation dynamics and crystal structure formations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Arginine is used in the synthesis of creatine and polyamines. DL-Arg is used in physicochemical analysis of amino acid complexation dynamics and crystal structure formations.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 7200-25-1
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.204
MDL Number MFCD00063117
InChI Key ODKSFYDXXFIFQN-UHFFFAOYSA-N
Synonym dl-arginine, arginin, h-dl-arg-oh, 2-amino-5-carbamimidamidopentanoic acid, 2-amino-5-guanidinopentanoic acid, unii-fl26ntk3ep, dl-arginine, free base, fl26ntk3ep, wln: muyzm3yzvq &gh, arginine, dl
PubChem CID 232
ChEBI CHEBI:29016
IUPAC Name 2-amino-5-(diaminomethylideneamino)pentanoic acid
SMILES C(CC(C(=O)O)N)CN=C(N)N

Specifications

Melting Point ∼230°C (decomposition)
Beilstein 1725411
Solubility Information Insoluble in water.
Quantity 25 g
Formula Weight 174.2
Sensitivity Air Sensitive
Percent Purity 98%
Chemical Name or Material DL-Arginine
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 230-571-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only