missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-Tocopherol Acetate 97.0+%, TCI America™

Catalog No. T025225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T025225G 25 g
T0252100G 100 g
T0252500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T025225G Supplier TCI America Supplier No. T025225G

Chemical Identifiers

CAS 7695-91-2
Molecular Formula C31H52O3
Molecular Weight (g/mol) 472.754
MDL Number MFCD00072042
InChI Key ZAKOWWREFLAJOT-CEFNRUSXSA-N
Synonym vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ
PubChem CID 86472
ChEBI CHEBI:32321
IUPAC Name [(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate
SMILES CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

Color Yellow
Boiling Point 224°C
Quantity 25 g
Formula Weight 472.75
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material DL-alpha-Tocopherol Acetate
EINECSNumber (9)-0487
RTECSNumber GA8747000
TSCA Yes