missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Alanine Methyl Ester Hydrochloride 98.0+%, TCI America™
$45.57 - $123.41
Chemical Identifiers
| CAS | 13515-97-4 |
|---|---|
| Molecular Formula | C4H10ClNO2 |
| Molecular Weight (g/mol) | 139.58 |
| MDL Number | MFCD00035523 |
| InChI Key | IYUKFAFDFHZKPI-UHFFFAOYNA-N |
| Synonym | methyl 2-aminopropanoate hydrochloride, dl-alanine methyl ester hydrochloride, h-dl-ala-ome.hcl, h-dl-ala-ome hydrochloride, dl-alanine methyl ester hcl, methyl l-alaninate hcl, methyl dl-alaninate hcl, methyl dl-alaninate hydrochloride, h-dl-ala-ome hcl, methyl dl-2-aminopropanoate hydrochloride |
| PubChem CID | 2756298 |
| IUPAC Name | methyl 2-aminopropanoate hydrochloride |
| SMILES | Cl.COC(=O)C(C)N |
Chemical Identifiers
| 13515-97-4 | |
| 139.58 | |
| IYUKFAFDFHZKPI-UHFFFAOYNA-N | |
| 2756298 | |
| Cl.COC(=O)C(C)N |
| C4H10ClNO2 | |
| MFCD00035523 | |
| methyl 2-aminopropanoate hydrochloride, dl-alanine methyl ester hydrochloride, h-dl-ala-ome.hcl, h-dl-ala-ome hydrochloride, dl-alanine methyl ester hcl, methyl l-alaninate hcl, methyl dl-alaninate hcl, methyl dl-alaninate hydrochloride, h-dl-ala-ome hcl, methyl dl-2-aminopropanoate hydrochloride | |
| methyl 2-aminopropanoate hydrochloride |
Specifications
| 158°C | |
| White-Yellow | |
| MFCD00035523 | |
| IYUKFAFDFHZKPI-UHFFFAOYNA-N | |
| methyl 2-aminopropanoate hydrochloride | |
| 2756298 | |
| ≥98.0% (HPLC,N) | |
| DL-Alanine Methyl Ester Hydrochloride |
| 13515-97-4 | |
| C4H10ClNO2 | |
| methyl 2-aminopropanoate hydrochloride, dl-alanine methyl ester hydrochloride, h-dl-ala-ome.hcl, h-dl-ala-ome hydrochloride, dl-alanine methyl ester hcl, methyl l-alaninate hcl, methyl dl-alaninate hcl, methyl dl-alaninate hydrochloride, h-dl-ala-ome hcl, methyl dl-2-aminopropanoate hydrochloride | |
| Cl.COC(=O)C(C)N | |
| 139.58 | |
| 139.58 | |
| Crystalline Powder |
Safety and Handling
TSCA : No