accessibility menu, dialog, popup

UserName

CH3OOH3CNH2HCl

CAS RN 13515-97-4

IUPAC Name: methyl 2-aminopropanoate hydrochloride
Synonyms: methyl 2-aminopropanoate hydrochloride
Molecular Weight (g/mol): 139.58
Molecular Formula: C4H10ClNO2
InChi Key: IYUKFAFDFHZKPI-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(C)N
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)