missing translation for 'onlineSavingsMsg'
Learn More

DL-Alanine Methyl Ester Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America A244625G

Quantity 25 g

 View more versions of this product

Catalog No. A244625G


Description

Description

Specifications

Chemical Identifiers

13515-97-4
139.58
IYUKFAFDFHZKPI-UHFFFAOYNA-N
2756298
Cl.COC(=O)C(C)N
C4H10ClNO2
MFCD00035523
methyl 2-aminopropanoate hydrochloride, dl-alanine methyl ester hydrochloride, h-dl-ala-ome.hcl, h-dl-ala-ome hydrochloride, dl-alanine methyl ester hcl, methyl l-alaninate hcl, methyl dl-alaninate hcl, methyl dl-alaninate hydrochloride, h-dl-ala-ome hcl, methyl dl-2-aminopropanoate hydrochloride
methyl 2-aminopropanoate hydrochloride

Specifications

158°C
13515-97-4
C4H10ClNO2
methyl 2-aminopropanoate hydrochloride, dl-alanine methyl ester hydrochloride, h-dl-ala-ome.hcl, h-dl-ala-ome hydrochloride, dl-alanine methyl ester hcl, methyl l-alaninate hcl, methyl dl-alaninate hcl, methyl dl-alaninate hydrochloride, h-dl-ala-ome hcl, methyl dl-2-aminopropanoate hydrochloride
Cl.COC(=O)C(C)N
25 g
2756298
≥98.0% (HPLC,N)
DL-Alanine Methyl Ester Hydrochloride
White-Yellow
MFCD00035523
IYUKFAFDFHZKPI-UHFFFAOYNA-N
methyl 2-aminopropanoate hydrochloride
139.58
139.58
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions