missing translation for 'onlineSavingsMsg'
Learn More

DL-Alanine Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. A24465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24465G 5 g
A244625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24465G Supplier TCI America Supplier No. A24465G

Chemical Identifiers

CAS 13515-97-4
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00035523
InChI Key IYUKFAFDFHZKPI-UHFFFAOYNA-N
Synonym methyl 2-aminopropanoate hydrochloride, dl-alanine methyl ester hydrochloride, h-dl-ala-ome.hcl, h-dl-ala-ome hydrochloride, dl-alanine methyl ester hcl, methyl l-alaninate hcl, methyl dl-alaninate hcl, methyl dl-alaninate hydrochloride, h-dl-ala-ome hcl, methyl dl-2-aminopropanoate hydrochloride
PubChem CID 2756298
IUPAC Name methyl 2-aminopropanoate hydrochloride
SMILES Cl.COC(=O)C(C)N

Specifications

Melting Point 158°C
Color White-Yellow
Quantity 5 g
Formula Weight 139.58
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material DL-Alanine Methyl Ester Hydrochloride