missing translation for 'onlineSavingsMsg'
Learn More

DL-Adrenaline Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America A090925G

 View more versions of this product

Catalog No. A090925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

DL-Adrenaline Hydrochloride
157°C
C9H14ClNO3
25 g
DL-Epinephrine Hydrochloride
CNCC(C1=CC(=C(C=C1)O)O)O.Cl
219.665
219.67
Crystalline Powder
329-63-5
White-Yellow
MFCD00050562
2811
ATADHKWKHYVBTJ-UHFFFAOYSA-N
4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol;hydrochloride
5924
≥98.0% (HPLC,T)

Chemical Identifiers

329-63-5
219.665
ATADHKWKHYVBTJ-UHFFFAOYSA-N
5924
CNCC(C1=CC(=C(C=C1)O)O)O.Cl
C9H14ClNO3
MFCD00050562
DL-Epinephrine Hydrochloride
4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : DO3330000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions