missing translation for 'onlineSavingsMsg'
Learn More

DL-Adrenaline Hydrochloride 98.0+%, TCI America™

Catalog No. p-7117125
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09095G 5 g
A090925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09095G Supplier TCI America Supplier No. A09095G
Only null left

Chemical Identifiers

CAS 329-63-5
Molecular Formula C9H14ClNO3
Molecular Weight (g/mol) 219.665
MDL Number MFCD00050562
InChI Key ATADHKWKHYVBTJ-UHFFFAOYSA-N
Synonym DL-Epinephrine Hydrochloride
PubChem CID 5924
IUPAC Name 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol;hydrochloride
SMILES CNCC(C1=CC(=C(C=C1)O)O)O.Cl

Specifications

Melting Point 157°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 219.67
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Adrenaline Hydrochloride
RTECSNumber DO3330000
TSCA Yes