missing translation for 'onlineSavingsMsg'
Learn More

DL-3-Hydroxybutyric Acid (contains Polymolecular esterification product) 80.0+%, TCI America™

Catalog No. H02285G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H02285G 5 g
H022825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H02285G Supplier TCI America Supplier No. H02285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 300-85-6
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00004546
InChI Key WHBMMWSBFZVSSR-UHFFFAOYSA-N
Synonym 3-hydroxybutyric acid, butanoic acid, 3-hydroxy, beta-hydroxybutyric acid, dl-beta-hydroxybutyric acid, beta-hydroxybuttersaeure, 3 hba, 3-hydroxybuttersaeure, beta-hydroxy-n-butyric acid, 3 hydroxybutyrate, butyric acid, 3-hydroxy
PubChem CID 441
ChEBI CHEBI:20067
IUPAC Name 3-hydroxybutanoic acid
SMILES CC(CC(=O)O)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 104.11
Percent Purity ≥80.0% (T)
Physical Form Liquid
Chemical Name or Material DL-3-Hydroxybutyric Acid (contains Polymolecular esterification product)
EINECSNumber (2)-1429
TSCA Yes
Recommended Storage Refrigerator