missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Methyl-1-butanol (Synthetic) 97.0+%, TCI America™

Catalog No. M017525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M017525ML 25 mL
M0175500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M017525ML Supplier TCI America Supplier No. M017525ML

Chemical Identifiers

CAS 137-32-6
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004743
InChI Key QPRQEDXDYOZYLA-UHFFFAOYSA-N
Synonym 2-methyl-1-butanol, active amyl alcohol, 1-butanol, 2-methyl, 2-methylbutanol, sec-butylcarbinol, dl-2-methyl-1-butanol, 2-methyl-n-butanol, 2-methylbutyl alcohol, 2-methyl butanol-1, primary active amyl alcohol
PubChem CID 8723
ChEBI CHEBI:48945
IUPAC Name 2-methylbutan-1-ol
SMILES CCC(C)CO

Specifications

Color Colorless
Boiling Point 128°C
Quantity 25 mL
UN Number 1105
Formula Weight 88.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material DL-2-Methyl-1-butanol (Synthetic)
EINECSNumber (2)-0217
RTECSNumber EL5250000
TSCA Yes