missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-1,4-Dithiothréitol, 99+%, pour la biologie moléculaire, sans ADN, ARNase et protéase

Numéro de catalogue. AC327190010
Modifier l'affichage
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Bouteille en plastique
Bouteille en verre
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC327190010 1 g Bouteille en verre
AC327190100 10 g Bouteille en plastique
AC327190500 50 g Bouteille en plastique
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC327190010 Fournisseur Thermo Scientific Chemicals Code fournisseur 327190010
Obtenez jusqu’à 1,800 $ en produits chimiques
Afficher toutes les promotions

CAS : 3483-12-3 | C4H10O2S2 | 154,24 g/mol

Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.

Identifiants chimiques

CAS 12-3-3483
ChEBI CHEBI:42170

Spécifications

Absorbance (0.1 M in water), (0.1 M in water) at 0nm, 0.1 max. at 280nm, 0.5 max. at 260nm
Melting Point 39.0°C to 44.0°C
Color Incolore à jaune
Boiling Point 125.0°C to 130.0°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Bouteille en verre
Molecular Formula C4H10O2S2
Linear Formula HSCH2CH(OH)CH(OH)CH2SH
Quantity 1 g
MDL Number MFCD00004877
Beilstein 01, III, 2360
Merck Index 15, 3419
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
SMILES C(C(C(CS)O)O)S
IUPAC Name (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
Molecular Weight (g/mol) 154.24
PubChem CID 446094
Formula Weight 154.24
Percent Purity ≥99%
Grade Biologie moléculaire
Physical Form Poudre
Chemical Name or Material DL-1, 4-Dithiothreitol, DNase, RNase, Protease free
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 248-531-9
RTECSNumber XO8576500

RUO – Usage réservé à la recherche