missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-1,4-Dithiothreitol, 99%, for biochemistry
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
- 1,4-Dithiothreitol (DTT or Cleland's reagent) reduces disulfide bonds in proteins, leading to loss of native conformation
- DTT is not stable in solution and should be prepared in small aliquots prior to use; the use of EDTA in the solution can increase the half-life of DTT
- Typically used at a final concentration of 0.1M for protein stabilization; recommended to create a 1M stock
Applications
- Protects thiol groups from oxidation and acts as a chemical chelator in solution; maintains proteins in the denatured state
- Common applications include SDS-page electrophoresis and protein isolation
Spécifications
Spécifications
| Absorbance | (1 M soln. in water), (1 M soln. in water) at 0nm, 0.035 max. at 500nm |
| Melting Point | 39°C to 44°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 0.99 |
| Molecular Formula | C4H10O2S2 |
| Linear Formula | HSCH2CH(OH)CH(OH)CH2SH |
| Quantity | 25 g |
| Beilstein | 01, III, 2360 |
| Merck Index | 15, 3419 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2) |
| Hazard Statement | Causes serious eye irritation, Harmful if swallowed, May cause respiratory irritation, May form combustible dust concentrations in air, Causes skin irritation |
RUO – Research Use Only