missing translation for 'onlineSavingsMsg'
Learn More

Dithiooxamide, 98%

Catalog No. AC117551000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC117551000 100 g
AC117550050 5 g
AC117550250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC117551000 Supplier Thermo Scientific Chemicals Supplier No. 117551000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79-40-3 | C2H4N2S2 | 120.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-40-3
Molecular Formula C2H4N2S2
Molecular Weight (g/mol) 120.19
InChI Key OAEGRYMCJYIXQT-UHFFFAOYSA-N
Synonym dithiooxamide, rubeanic acid, ethanebis thioamide, oxamide, dithio, dithioxamide, rubeane, rubean, hydrorubeanic acid, oxaldiimidic acid, dithio, usaf mk-6
PubChem CID 2777982
IUPAC Name ethanedithioamide
SMILES C(=S)(C(=S)N)N

Specifications

Melting Point 200.0°C
Elemental Analysis 22.7 to 23.9% (N) 19.5 to 20.5% (C)
Infrared Spectrum Authentic
Linear Formula H2NCSCSNH2
Quantity 100 g
Merck Index 15,8412
Solubility Information Solubility in water: 0.37g/L (20°C).
Formula Weight 120.19
Percent Purity 98%
Chemical Name or Material Dithiooxamide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 201-203-9

RUO – Research Use Only