Learn More
Abcam Inc Disuccinimidyl suberate, cross-linking reagent, 500MG
Numéro de catalogue. 502706726
Encompass
MW 368.34 Da, Purity >99%. Homobifunctional primary amine reactive cross-linker. Contains an 8 atom linker (11.4 ) and can cross-link distances up to ~25 due to protein flexibility. Produces no toxic side products.
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502706726 Fournisseur Abcam Inc Code fournisseur AB141274500MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
MW 368.34 Da, Purity >99%. Homobifunctional primary amine reactive cross-linker. Contains an 8 atom linker (11.4 ) and can cross-link distances up to ~25 due to protein flexibility. Produces no toxic side products.
The product is subject to the following: Abcam Restricted Use Statement