Learn More
Waters Limited DisQuE AOAC Method Sample Preparation Kit, Tubes
Numéro de catalogue. 50780055
Encompass
DisQuE Dispersive Sample Preparation used for QUECHERS Method AOAC Method 2007.01. The kit consists of 100 units each of extraction tubes and clean-up tubes. Each 50 mL DisQuE Extraction tube contains 6 grams of magnesium sulphate and 1.5 grams of sodium acetate. Each 2 mL DisQuE Clean-Up tube contains 150 mg of magnesium sulphate and 50 mg PSA (primary-secondary amine SPE sorbent). Detailed method instructions are included with the kit. QUECHERS (pronounced catchers) methods are used for multi-pesticide residue analyses in food.
Numéro de catalogue. 50780055 Fournisseur Waters Limited Code fournisseur 176001676
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
DisQuE Dispersive Sample Preparation used for QUECHERS Method AOAC Method 2007.01. The kit consists of 100 units each of extraction tubes and clean-up tubes. Each 50 mL DisQuE Extraction tube contains 6 grams of magnesium sulphate and 1.5 grams of sodium acetate. Each 2 mL DisQuE Clean-Up tube contains 150 mg of magnesium sulphate and 50 mg PSA (primary-secondary amine SPE sorbent). Detailed method instructions are included with the kit. QUECHERS (pronounced catchers) methods are used for multi-pesticide residue analyses in food.