Learn More
Research Products Corporation Disposable Universal Electroporation Cuvettes, 1 mm Gap, 100 UL Capacity, White Square Lid, 50 per Case
Numéro de catalogue. 501537657
Encompass
Disposable Universal Electroporation Cuvettes are designed for use with bacterial, yeast, fungi, and mammalian cells. Cuvettes are manufactured from the clearest, highest quality, durable, polycarbonate material and have demonstrated optimal transfection efficiencies over decades of laboratory use. Compatible with any Electroporators or devices which require the use of electroporation cuvettes (Eporator, BioRad, Gene Pulsar and BTX).
Precision built to give exacting tolerances and ensure reproducible results. Cuvettes are skillfully crafted and polished aluminum plates reduce static and arcing, delivering optimum performance. Individually wrapped and gamma irradiated to guarantee sterility. Individually color-coded cuvette lids identify application, with corresponding gap widths.
Numéro de catalogue. 501537657 Fournisseur Research Products Corporation Code fournisseur 212633
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Disposable Universal Electroporation Cuvettes are designed for use with bacterial, yeast, fungi, and mammalian cells. Cuvettes are manufactured from the clearest, highest quality, durable, polycarbonate material and have demonstrated optimal transfection efficiencies over decades of laboratory use. Compatible with any Electroporators or devices which require the use of electroporation cuvettes (Eporator, BioRad, Gene Pulsar and BTX).
Precision built to give exacting tolerances and ensure reproducible results. Cuvettes are skillfully crafted and polished aluminum plates reduce static and arcing, delivering optimum performance. Individually wrapped and gamma irradiated to guarantee sterility. Individually color-coded cuvette lids identify application, with corresponding gap widths.