missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disodium L-(+)-Tartrate Dihydrate 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Blanc |
| Quantity | 25 g |
| Formula Weight | 194.05 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | Disodium L-(+)-Tartrate Dihydrate |
Sécurité et manipulation
| TSCA | Yes |