missing translation for 'onlineSavingsMsg'
Learn More

Disodium Citrate Sesquihydrate 98.0+%, TCI America™

Catalog No. C0365500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0365500G 500 g
C036525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0365500G Supplier TCI America Supplier No. C0365500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5-4-6132
Molecular Formula C12H18Na4O17
Molecular Weight (g/mol) 526.22
MDL Number MFCD00150445
InChI Key HGPVLOQNBSHYEI-UHFFFAOYNA-J
Synonym unii-cz1032cekr, cz1032cekr, sodium citrate dibasic sesquihydrate, disodium citrate sesquihydrate, disodium hydrogen citrate 1.5 h2o, sodium hydrogen citrate sesquihydrate, citric acid, disodium salt, sesquihydrate, tetrasodium, 3-carboxy-3-hydroxypentanedioate, trihydrate, sodium citrate dibasic sesquihydrate, purum p.a t, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, disodium salt, hydrate 2:3
PubChem CID 117063816
IUPAC Name tetrasodium;2-(carboxymethyl)-2-hydroxybutanedioate;trihydrate
SMILES C(C(=O)O)C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)O)C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.[Na+].[Na+].[Na+].[Na+]

Specifications

Melting Point 149°C
Color White
pH 4.7 to 5.2 (50g/L aq. soln.)
Quantity 500 g
Formula Weight 236.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Citrate Sesquihydrate
EINECSNumber (2)-1323
RTECSNumber GE7580000
TSCA Yes