missing translation for 'onlineSavingsMsg'
Learn More

Disodium 4-Nitrophenyl Phosphate Hexahydrate 98.0+%, TCI America™

Catalog No. D40055G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40055G 5 g
D40051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40055G Supplier TCI America Supplier No. D40055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 333338-18-4
Molecular Formula C6H16NNa2O12P
Molecular Weight (g/mol) 371.142
MDL Number MFCD00066288
InChI Key KAKKHKRHCKCAGH-UHFFFAOYSA-L
Synonym Phosphoric Acid 4-Nitrophenyl Ester Disodium Salt
PubChem CID 2723710
IUPAC Name disodium;(4-nitrophenyl) phosphate;hexahydrate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OP(=O)([O-])[O-].O.O.O.O.O.O.[Na+].[Na+]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 263.05
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 4-Nitrophenyl Phosphate Hexahydrate [for Biochemical Research]
EINECSNumber (9)-0171
TSCA Yes
Recommended Storage Freezer