missing translation for 'onlineSavingsMsg'
Learn More

Disodium 1-Naphthyl Phosphate Hydrate [Substrate for Phosphatase], TCI America™

Catalog No. P02631G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P02631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P02631G Supplier TCI America Supplier No. P02631G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 207569-06-0
Molecular Formula C10H7Na2O4P
Molecular Weight (g/mol) 268.12
MDL Number MFCD00041007
InChI Key QYURIFWAOPAPAJ-UHFFFAOYSA-L
Synonym 1-naphthyl phosphate disodium salt hydrate, sodium naphthalen-1-yl phosphate hydrate, disodium 1-naphthyl phosphate hydrate, disodium hydrate naphthalen-1-yl phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h7o4p.2na.h2o, 1-naphthyl?phosphate?disodium?salt?hydrate, 1-naphthyl phosphate disodium salt monohydrate, dihydrogen naphthyl phosphate, oxamethane, sodium salt, sodium salt
PubChem CID 24212014
IUPAC Name disodium naphthalen-1-yl phosphate
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1

Specifications

Quantity 1 g
Formula Weight 268.12
Physical Form Crystalline Powder
Chemical Name or Material Disodium 1-Naphthyl Phosphate Hydrate [Substrate for Phosphatase]
TSCA No
Recommended Storage Freezer