missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dirithromycin, ≥95% (TLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 62013-04-1 |
| Physical Form | Solid |
| Linear Formula | C42H78N2O14 |
| MDL Number | MFCD00865041 |
| Synonym | [9 S(R)]-9-Deoxo-11-deoxy-9,11-[imino[2-(2-methoxy)ethylidene]oxy]erthromycin; LY-237216 |
| Type | Analytical Standard |
| Industry Type | Drug Analysis |
| Quantity | 1 g |
| Formula Weight | 835.07 |
| Percent Purity | ≥95% (TLC) |
| Show More |