missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DIRC2 Polyclonal Antibody

Description
Immunogen sequence: IPISDLILKRR LIHGGQMLNG LAGPTVMNAA PFLSTTWFSA DERATATAIA S Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.
Electrogenic metabolite transporter.
Spécifications
Spécifications
| Antigen | DIRC2 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Dirc2 |
| Gene Accession No. | Q96SL1 |
| Gene Alias | Dirc2; Disrupted in renal cancer protein 2; disrupted in renal carcinoma 2; disrupted in renal carcinoma 2 (human); disrupted in renal carcinoma 2 homolog; disrupted in renal carcinoma protein 2; Disrupted in renal carcinoma protein 2 homolog; RCC4; renal cell carcinoma 4; SLC49A4; Solute carrier family 49 member 4 |
| Gene Symbols | Dirc2 |
| Afficher plus de résultats |