missing translation for 'onlineSavingsMsg'
Learn More

Dipyridamole

Catalog No. p-7042677
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6332906 5 g
AAJ6332914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6332906 Supplier Thermo Scientific Chemicals Supplier No. J6332906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An inhibitor of adenosine transport and phosphodiesterase | CAS: 58-32-2 | C24H40N8O4 | 504.636 g/mol

Dipyridamole prevents platelets sticking to the replacement heart valve and causing a blood clot on the valve. It is used to dilate blood vessels in people with peripheral arterial disease and coronary artery disease. This compound has been shown to suppress high glucose-induced osteopontin mRNA expression and protein secretion, as well as inhibit cAMP and cGMP hydrolysis. Research indicates that Dipyridamole is a non-specific nucleoside transport inhibitor with the ability to increase the effects of adenosine in sinoatrial and atrioventricular nodes. Dipyridamole is an inhibitor of ENT1 and ENT2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dipyridamole prevents platelets sticking to the replacement heart valve and causing a blood clot on the valve. It is used to dilate blood vessels in people with peripheral arterial disease and coronary artery disease. This compound has been shown to suppress high glucose-induced osteopontin mRNA expression and protein secretion, as well as inhibit cAMP and cGMP hydrolysis. Research indicates that Dipyridamole is a non-specific nucleoside transport inhibitor with the ability to increase the effects of adenosine in sinoatrial and atrioventricular nodes. Dipyridamole is an inhibitor of ENT1 and ENT2.

Solubility
Soluble in chloroform, methanol, dilute acids, ethanol. Slightly soluble in water.

Notes
Store in a cool dry place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 58-32-2
Molecular Formula C24H40N8O4
Molecular Weight (g/mol) 504.636
MDL Number MFCD00010555
InChI Key IZEKFCXSFNUWAM-UHFFFAOYSA-N
Synonym dipyridamole, dipyridamine, persantin, dipyridamol, dipyudamine, persantine, curantyl, stimolcardio, cardoxin, kurantil
PubChem CID 3108
ChEBI CHEBI:4653
IUPAC Name 2-[[2-[bis(2-hydroxyethyl)amino]-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-6-yl]-(2-hydroxyethyl)amino]ethanol
SMILES C1CCN(CC1)C2=NC(=NC3=C2N=C(N=C3N4CCCCC4)N(CCO)CCO)N(CCO)CCO

Specifications

Melting Point 165°C to 167°C
Color Yellow
Quantity 5 g
Merck Index 14,3346
Solubility Information Soluble in chloroform,methanol,dilute acids,ethanol. Slightly soluble in water.
Formula Weight 504.63
Physical Form Powder
Chemical Name or Material Dipyridamole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-374-7
RTECSNumber KK7450000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only