missing translation for 'onlineSavingsMsg'
Learn More

Dipyridamole 98.0+%, TCI America™

Catalog No. p-7127209
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22745G 5 g
D227425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22745G Supplier TCI America Supplier No. D22745G

Chemical Identifiers

CAS 58-32-2
Molecular Formula C24H40N8O4
Molecular Weight (g/mol) 504.636
MDL Number MFCD00010555
InChI Key IZEKFCXSFNUWAM-UHFFFAOYSA-N
Synonym dipyridamole, dipyridamine, persantin, dipyridamol, dipyudamine, persantine, curantyl, stimolcardio, cardoxin, kurantil
PubChem CID 3108
ChEBI CHEBI:4653
IUPAC Name 2-[[2-[bis(2-hydroxyethyl)amino]-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-6-yl]-(2-hydroxyethyl)amino]ethanol
SMILES C1CCN(CC1)C2=NC(=NC3=C2N=C(N=C3N4CCCCC4)N(CCO)CCO)N(CCO)CCO

Specifications

Melting Point 168°C
Color Yellow
Quantity 5 g
Formula Weight 504.64
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dipyridamole
RTECSNumber KK7450000
TSCA No