missing translation for 'onlineSavingsMsg'
Learn More

Dipropylamine 99.0+%, TCI America™

Catalog No. D093025ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D093025ML 25 mL
D0930100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D093025ML Supplier TCI America Supplier No. D093025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142-84-7
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00009362
InChI Key WEHWNAOGRSTTBQ-UHFFFAOYSA-N
Synonym dipropylamine, di-n-propylamine, 1-propanamine, n-propyl, n-dipropylamine, n-propyl-1-propanamine, rcra waste number u110, dipropyl amine, rcra waste no. u110, unii-60p318iiry, ccris 4805
PubChem CID 8902
IUPAC Name dipropylamine
SMILES CCCNCCC

Specifications

Melting Point -63°C
Color Yellow
Boiling Point 108°C
Quantity 25 mL
UN Number 2383
Formula Weight 101.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Dipropylamine
EINECSNumber (2)-3086
RTECSNumber JL9200000
TSCA Yes