missing translation for 'onlineSavingsMsg'
Learn More

Dipropyl Disulfide 98.0+%, TCI America™

Catalog No. D0932500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0932500ML 500 mL
D093225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0932500ML Supplier TCI America Supplier No. D0932500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 629-19-6
Molecular Formula C6H14S2
Molecular Weight (g/mol) 150.30
MDL Number MFCD00009378
InChI Key ALVPFGSHPUPROW-UHFFFAOYSA-N
Synonym dipropyl disulfide, propyl disulfide, disulfide, dipropyl, n-propyl disulfide, di-n-propyl disulfide, 4,5-dithiaoctane, dipropyl disulphide, 1-propyldisulfanyl propane, propyldithiopropane, 1,1'-dithiodipropane
PubChem CID 12377
ChEBI CHEBI:45758
IUPAC Name 1-(propyldisulfanyl)propane
SMILES CCCSSCCC

Specifications

Color Yellow
Boiling Point 193°C
Quantity 500 mL
Formula Weight 150.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dipropyl Disulfide
EINECSNumber (2)-0477&(2)-2421
RTECSNumber JO1955000
TSCA Yes