missing translation for 'onlineSavingsMsg'
Learn More

Dipotassium Glycyrrhizinate Hydrate 75.0+%, TCI America™

Catalog No. G027025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G027025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G027025G Supplier TCI America Supplier No. G027025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68797-35-3
Molecular Formula C42H62K2O17
Molecular Weight (g/mol) 917.138
MDL Number MFCD02094742
InChI Key MWDRYYYKIRPGSC-DWJAGBRCSA-L
Synonym Glycyrrhizic Acid Dipotassium Salt, Glycyrrhizin Dipotassium Salt
PubChem CID 91641377
IUPAC Name dipotassium;(2S,3S,4S,5R,6R)-6-[(2S,3R,4S,5S,6S)-2-[[(3S,4aR,6aR,6bS,8aS,11S,12aR,14aR,14bS)-11-carboxy-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-2,3,4a,5,6,7,8,9,10,12,12a,14a-dodecahydro-1H-picen-3-yl]oxy]-6-carboxylato-4,5-dihydroxyoxan-3-yl]oxy-3,4,5-tri
SMILES CC1(C2CCC3(C(C2(CCC1OC4C(C(C(C(O4)C(=O)[O-])O)O)OC5C(C(C(C(O5)C(=O)[O-])O)O)O)C)C(=O)C=C6C3(CCC7(C6CC(CC7)(C)C(=O)O)C)C)C)C.O.[K+].[K+]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 899.12
Percent Purity ≥75.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dipotassium Glycyrrhizinate Hydrate
EINECSNumber (8)-0502
TSCA Yes