missing translation for 'onlineSavingsMsg'
Learn More

Diphenylsilanediol, 98%

Catalog No. AAA1049222
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1049222 100 g
AAA1049236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1049222 Supplier Thermo Scientific Chemicals Supplier No. A1049222

CAS: 947-42-2 | C12H12O2Si | 216.311 g/mol

Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.

Solubility
Reacts with water.

Notes
Stable under normal temperatures and pressures. Incompatible with Oxidizing agents. Store at room temperature. Air & Light Sensitive.

Chemical Identifiers

CAS 947-42-2
Molecular Formula C12H12O2Si
Molecular Weight (g/mol) 216.311
MDL Number MFCD00002101
InChI Key OLLFKUHHDPMQFR-UHFFFAOYSA-N
Synonym diphenylsilanediol, silanediol, diphenyl, diphenyldihydroxysilane, difenyl-dihydroxysilan, silane, dihydroxydiphenyl, dihydroxy diphenyl silane, diphenylsilandiol, silanediol, 1,1-diphenyl, unii-36x37p8nbb, difenyl-dihydroxysilan czech
PubChem CID 13693
IUPAC Name dihydroxy(diphenyl)silane
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(O)O

Specifications

Melting Point ∼150°C (decomposition)
Quantity 100 g
Linear Formula (C6H5Si(OH)2
UN Number UN1325
Beilstein 2523445
Solubility Information Reacts with water.
Formula Weight 216.31
Percent Purity 98%
Odor Odorless
Chemical Name or Material Diphenylsilanediol
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 213-427-4
RTECSNumber VV3640000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only