missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphine oxide, 97%

Catalog No. AC310120010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC310120010 1 g Glass bottle
AC310120050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC310120010 Supplier Thermo Scientific Chemicals Supplier No. 310120010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4559-70-0 | C12H11OP | 202.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4559-70-0
Molecular Formula C12H11OP
Molecular Weight (g/mol) 202.19
MDL Number MFCD00002079
InChI Key ASUOLLHGALPRFK-UHFFFAOYSA-N
Synonym diphenylphosphine oxide, phosphine oxide, diphenyl, diphenyl phosphine oxide, hpoph2, asuollhgalprfk-uhfffaoysa-n, dppo, diphenylphosphane oxide, phenylphosphonoylbenzene, diphenylphosphino-1-one, pubchem23810
PubChem CID 6327869
IUPAC Name oxo(diphenyl)phosphanium
SMILES O=P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 50.0°C to 57.0°C
Color Orange to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6080 to 1.6100
Linear Formula (C6H5)2P(O)H
Quantity 1 g
Formula Weight 202.2
Percent Purity 97%
Physical Form Crystals
Chemical Name or Material Diphenylphosphine oxide
Show More Show Less

RUO – Research Use Only