missing translation for 'onlineSavingsMsg'
Learn More

Diphenylmethanol, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022864
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022864 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022864 Supplier Merck Emd Millipore Supplier No. BP1119

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 91-01-0
Melting Point 65°C to 67°C (literature)
Boiling Point 297°C to 298°C (literature)
Molecular Formula C13H12O
Linear Formula (C6H5)2 CHOH
MDL Number MFCD00004488
Quantity 25 mg
Synonym Benzhydrol; Benzhydryl alcohol; Diphenyl carbinol
Formula Weight 184.23
Grade BP
Show More Show Less